Search
Search

LARS1| Gene Symbol: LARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102492.1 | NP_001095962.1 | 18 - 19 | 1874 | 69 | |
| GenBank mRNA | BC148056.1 | - | 18 - 19 | 1874 | 69 | |
Set Membership: |
|