Search
Search

PRDM8| Gene Symbol: PRDM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103232.1 | NP_001096702.1 | 1 - 2 | 240 | 85 | |
| GenBank mRNA | BC151684.1 | - | 1 - 2 | 240 | 85 | |
| BC151777.1 | - | 1 - 2 | 241 | 85 | ||
Set Membership: |
|