Search
Search

ANO10| Gene Symbol: ANO10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102169.1 | NP_001095639.1 | 10 - 11 | 1775 | 104 | |
| GenBank mRNA | BC151532.1 | - | 10 - 11 | 1775 | 104 | |
Set Membership: |
|