Search
Search
| Gene Symbol: EBD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175703.3 | NP_783634.1 | 2 - 3 | 538 | 91 | |
| GenBank mRNA | AF000362.1 | - | 1 - 2 | 83 | 91 | |
| BC114788.1 | - | 2 - 3 | 538 | 91 | ||
Set Membership: |
|