Search
Search
| Gene Symbol: PGD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143738.1 | NP_001137210.1 | 11 - 12 | 1297 | 100 | |
| GenBank mRNA | AB098990.1 | - | 2 - 3 | 185 | 100 | |
| BC102178.1 | - | 11 - 12 | 1297 | 100 | ||
Set Membership: |
|