Search
Search

CDK13| Gene Symbol: CDK13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205431.1 | NP_001192360.1 | 2 - 3 | 1880 | 100 | |
| NM_001206622.1 | NP_001193551.1 | 2 - 3 | 1880 | 100 | ||
| GenBank mRNA | DV800787.1 | - | 1 - 2 | 385 | 100 | |
Set Membership: |
|