Search
Search

NDUFAF5| Gene Symbol: NDUFAF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206395.1 | NP_001193324.1 | 8 - 9 | 813 | 112 | |
| GenBank mRNA | BC122720.1 | - | 8 - 9 | 814 | 112 | |
Set Membership: |
|