Search
Search

ser-2| Gene Symbol: ser-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029164.3 | NP_001024335.1 | 6 - 7 | 644 | 59 | |
| NM_001029165.5 | NP_001024336.1 | 6 - 7 | 1120 | 59 | ||
| NM_001029168.4 | NP_001024339.1 | 6 - 7 | 644 | 59 | ||
| NM_001029169.5 | NP_001024340.1 | 5 - 6 | 729 | 59 | ||
| NM_001270331.4 | NP_001257260.1 | 6 - 7 | 644 | 59 | ||
| GenBank mRNA | GQ217520.1 | - | 5 - 6 | 611 | 59 | |
Set Membership: |
|