Search
Search

nkat-3| Gene Symbol: nkat-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029651.5 | NP_001024822.1 | 2 - 3 | 443 | 116 | |
| NM_001029652.6 | NP_001024823.1 | 2 - 3 | 371 | 116 | ||
Set Membership: |
|