Search
Search

ctn-1| Gene Symbol: ctn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001377804.1 | NP_001364770.1 | 14 - 15 | 2421 | 67 | |
| NM_001381436.2 | NP_001367838.1 | 13 - 14 | 2386 | 67 | ||
| NR_131363.1 | 13 - 14 | 2356 | 67 | |||
Set Membership: |
|