Search
Search

wrk-1| Gene Symbol: wrk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029476.4 | NP_001024647.1 | 6 - 7 | 751 | 70 | |
| NM_001029477.5 | NP_001024648.1 | 7 - 8 | 779 | 70 | ||
| NM_001029480.6 | NP_001024651.1 | 7 - 8 | 797 | 70 | ||
| NM_001373391.4 | NP_001360746.1 | 7 - 8 | 786 | 70 | ||
Set Membership: |
|