Search
Search

nhr-71| Gene Symbol: nhr-71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029616.6 | NP_001024787.1 | 5 - 6 | 1005 | 99 | |
| NM_001029617.5 | NP_001024788.1 | 5 - 6 | 882 | 99 | ||
Set Membership: |
|