Search
Search

gei-4| Gene Symbol: gei-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027640.5 | NP_001022811.1 | 5 - 6 | 888 | 80 | |
| NM_001381690.1 | NP_001367960.1 | 8 - 9 | 1189 | 80 | ||
| NM_001384004.1 | 24 - 25 | 3659 | 80 | |||
| NM_001384005.1 | 6 - 7 | 947 | 80 | |||
| NM_001384006.1 | 4 - 5 | 1305 | 80 | |||
| NM_064789.5 | NP_497190.1 | 10 - 11 | 1691 | 80 | ||
Set Membership: |
|