Search
Search

| Gene Symbol: srm-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074154.5 | NP_506555.2 | 1 - 2 | 318 | 121 | |
| NM_182385.4 | NP_872185.2 | 1 - 2 | 224 | 121 | ||
| Gene Symbol: srm-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028771.4 | NP_001023942.1 | 6 - 7 | 1665 | 121 | |
Set Membership: |
|