Search
Search

shn-1| Gene Symbol: shn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026835.6 | NP_001022006.1 | 1 - 2 | 138 | 82 | |
| NM_001026836.6 | NP_001022007.1 | 1 - 2 | 241 | 82 | ||
| NM_001383954.1 | 1 - 2 | 140 | 82 | |||
Set Membership: |
|