Search
Search

dex-1| Gene Symbol: dex-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379713.1 | NP_001367708.1 | 12 - 13 | 3041 | 79 | |
| NM_065780.6 | NP_498181.3 | 11 - 12 | 2965 | 79 | ||
| NM_065781.6 | NP_498182.2 | 12 - 13 | 3008 | 79 | ||
Set Membership: |
|