Search
Search

col-99| Gene Symbol: col-99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129303.4 | NP_001122775.2 | 7 - 8 | 597 | 104 | |
| NM_001307633.3 | NP_001294562.1 | 6 - 7 | 570 | 104 | ||
| NM_001307634.4 | NP_001294563.1 | 6 - 7 | 570 | 104 | ||
| NM_001322768.3 | NP_001309611.1 | 6 - 7 | 570 | 104 | ||
| NM_001322769.3 | NP_001309607.1 | 6 - 7 | 570 | 104 | ||
| NM_001322770.3 | NP_001309609.1 | 6 - 7 | 570 | 104 | ||
| NM_001322771.3 | NP_001309605.1 | 6 - 7 | 570 | 104 | ||
| NM_001322775.2 | NP_001309601.1 | 6 - 7 | 570 | 104 | ||
| NM_067468.7 | NP_499869.3 | 6 - 7 | 570 | 104 | ||
| GenBank mRNA | KM875546.1 | - | 6 - 7 | 570 | 104 | |
Set Membership: |
|