Search
Search

csp-2| Gene Symbol: csp-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380264.1 | NP_001368632.1 | 6 - 7 | 1248 | 90 | |
| NM_001383103.1 | 9 - 10 | 1550 | 90 | |||
| GenBank mRNA | AF088288.1 | - | 9 - 10 | 1547 | 90 | |
Set Membership: |
|