Search
Search

| Gene Symbol: T25F10.4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072309.3 | NP_504710.1 | 6 - 7 | 787 | 75 | |
| Gene Symbol: bbs-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072310.6 | NP_504711.2 | 13 - 14 | 2641 | 75 | |
Set Membership: |
|