Search
Search

hmp-1| Gene Symbol: hmp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129519.3 | NP_001122991.1 | 4 - 5 | 1579 | 87 | |
| NM_001276937.3 | NP_001263866.1 | 4 - 5 | 1578 | 87 | ||
| NM_001373121.3 | NP_001360176.1 | 5 | 1585 | 87 | ||
| NM_001383414.1 | 4 - 5 | 1579 | 87 | |||
| NM_073584.6 | NP_505985.1 | 4 - 5 | 1579 | 87 | ||
| GenBank mRNA | AF016852.1 | - | 4 - 5 | 1601 | 87 | |
Set Membership: |
|