Search
Search

gar-3| Gene Symbol: gar-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029064.4 | NP_001024235.1 | 9 - 10 | 1691 | 89 | |
| NM_001029065.5 | NP_001024236.1 | 8 - 9 | 1624 | 89 | ||
| GenBank mRNA | AF139093.2 | - | 9 - 10 | 1713 | 89 | |
| AY221631.1 | - | 8 - 9 | 1624 | 89 | ||
Set Membership: |
|