Search
Search

cng-1| Gene Symbol: cng-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129446.4 | NP_001122918.1 | 7 - 8 | 1705 | 125 | |
| NR_070019.1 | 8 - 9 | 1500 | 125 | |||
| NR_070020.1 | 7 - 8 | 1377 | 125 | |||
Set Membership: |
|