Search
Search

nhr-50| Gene Symbol: nhr-50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_074614.8 | NP_507015.3 | 5 - 6 | 888 | 130 | |
| NR_136352.1 | 3 - 4 | 769 | 130 | |||
| NR_136353.1 | 5 - 6 | 820 | 130 | |||
| NR_136354.1 | 5 - 6 | 844 | 130 | |||
Set Membership: |
|