Search
Search

CSNK1G3| Gene Symbol: CSNK1G3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626450.3 | XP_005626507.1 | 9 - 10 | 1351 | 126 | |
| XM_005626451.3 | XP_005626508.1 | 9 - 10 | 1696 | 126 | ||
| XM_014117683.2 | XP_013973158.1 | 9 - 10 | 1459 | 126 | ||
| XM_014117684.2 | XP_013973159.1 | 9 - 10 | 1092 | 126 | ||
| XM_022425217.1 | XP_022280925.1 | 9 - 10 | 1415 | 126 | ||
| XM_022425218.1 | XP_022280926.1 | 9 - 10 | 1693 | 126 | ||
| XM_022425219.1 | XP_022280927.1 | 9 - 10 | 1696 | 126 | ||
| XM_022425220.1 | XP_022280928.1 | 9 - 10 | 1696 | 126 | ||
| XM_022425221.1 | XP_022280929.1 | 9 - 10 | 1693 | 126 | ||
| XM_022425222.1 | XP_022280930.1 | 9 - 10 | 1696 | 126 | ||
| XM_022425223.1 | XP_022280931.1 | 9 - 10 | 1693 | 126 | ||
| XM_022425224.1 | XP_022280932.1 | 9 - 10 | 1696 | 126 | ||
| XM_022425225.1 | XP_022280933.1 | 10 - 11 | 1184 | 126 | ||
Set Membership: |
|