Search
Search
| Gene Symbol: RTBDN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024961.1 | NP_001020132.1 | 2 - 3 | 258 | 65 | |
| XM_005632322.2 | XP_005632379.1 | 3 - 4 | 543 | 65 | ||
| XM_014121734.2 | XP_013977209.1 | 4 - 5 | 2714 | 65 | ||
| XM_022406232.1 | XP_022261940.1 | 4 - 5 | 2344 | 65 | ||
| XM_022406233.1 | XP_022261941.1 | 2 - 3 | 1116 | 65 | ||
| GenBank mRNA | DQ058011.1 | - | 2 - 3 | 258 | 65 | |
Set Membership: |
|