Search
Search

TRAPPC12| Gene Symbol: TRAPPC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630090.3 | XP_005630147.1 | 2175 | 78 | ||
| XM_005630091.3 | XP_005630148.1 | 2488 | 78 | |||
| XM_022404401.1 | XP_022260109.1 | 2206 | 78 | |||
| XM_022404402.1 | XP_022260110.1 | 2169 | 78 | |||
| XM_532857.6 | XP_532857.2 | 2234 | 78 | |||
Set Membership: |
|