Search
Search

ZFYVE26| Gene Symbol: ZFYVE26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022422156.1 | XP_022277864.1 | 38 - 39 | 7209 | 112 | |
| XM_022422157.1 | XP_022277865.1 | 38 - 39 | 7311 | 112 | ||
| XM_022422158.1 | XP_022277866.1 | 38 - 39 | 7182 | 112 | ||
| XM_547867.6 | XP_547867.2 | 39 - 40 | 7380 | 112 | ||
Set Membership: |
|