Search
Search

PLEKHG4| Gene Symbol: PLEKHG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620823.3 | XP_005620880.1 | 15 - 16 | 2824 | 120 | |
| XM_014114094.2 | XP_013969569.1 | 15 - 16 | 2892 | 120 | ||
| XM_014114096.2 | XP_013969571.1 | 14 - 15 | 2939 | 120 | ||
| XM_022419321.1 | XP_022275029.1 | 15 - 16 | 2857 | 120 | ||
Set Membership: |
|