Search
Search
| Gene Symbol: EFNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622754.3 | XP_005622811.1 | 3 | 475 | 131 | |
| XM_005622755.3 | XP_005622812.1 | 3 | 758 | 131 | ||
| XM_005622756.3 | XP_005622813.1 | 3 | 630 | 131 | ||
| XM_014115357.2 | XP_013970832.1 | 3 | 618 | 131 | ||
| XM_846978.5 | XP_852071.1 | 3 | 566 | 131 | ||
Set Membership: |
|