Search
Search
| Gene Symbol: LRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640281.3 | XP_005640338.1 | 32 - 33 | 5594 | 158 | |
| XM_022414669.1 | XP_022270377.1 | 33 - 34 | 5609 | 158 | ||
| XM_022414670.1 | XP_022270378.1 | 33 - 34 | 5603 | 158 | ||
| XM_022414671.1 | XP_022270379.1 | 24 - 25 | 4363 | 158 | ||
Set Membership: |
|