Search
Search
| Gene Symbol: ARPP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638422.3 | XP_005638479.1 | 1 - 2 | 164 | 109 | |
| XM_022412585.1 | XP_022268293.1 | 2 - 3 | 158 | 109 | ||
| XM_022412586.1 | XP_022268294.1 | 2 - 3 | 147 | 109 | ||
Set Membership: |
|