Search
Search
| Gene Symbol: UPK1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022404366.1 | XP_022260074.1 | 4 - 5 | 468 | 63 | |
| XM_022404369.1 | XP_022260077.1 | 4 - 5 | 468 | 63 | ||
| XM_022404372.1 | XP_022260080.1 | 4 - 5 | 468 | 63 | ||
| XM_541693.4 | XP_541693.1 | 3 - 4 | 402 | 63 | ||
Set Membership: |
|