Search
Search
| Gene Symbol: TRAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003267.1 | NP_001003267.1 | 4 - 5 | 528 | 128 | |
| XM_014109166.2 | XP_013964641.1 | 4 - 5 | 517 | 128 | ||
| GenBank mRNA | X63678.1 | - | 528 | 128 | ||
Set Membership: |
|