Search
Search

Ccnt1| Gene Symbol: Ccnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515210.2 | XP_003515258.1 | 5 - 6 | 612 | 63 | |
| XM_007655202.1 | XP_007653392.1 | 5 - 6 | 610 | 63 | ||
| GenBank mRNA | JI879396.1 | - | 5 - 6 | 511 | 63 | |
| JP056727.1 | - | 5 - 6 | 510 | 63 | ||
Set Membership: |
|