Search
Search

Ipo13| Gene Symbol: Ipo13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502689.2 | XP_003502737.1 | 15 - 16 | 3168 | 95 | |
| GenBank mRNA | JI886584.1 | - | 14 - 15 | 1908 | 95 | |
| JP059471.1 | - | 14 - 15 | 1908 | 95 | ||
Set Membership: |
|