Search
Search

Osbpl2| Gene Symbol: Osbpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503110.2 | XP_003503158.1 | 10 - 11 | 1192 | 63 | |
| XM_007643738.1 | XP_007641928.1 | 11 - 12 | 1305 | 63 | ||
| XM_007643739.1 | XP_007641929.1 | 10 - 11 | 1191 | 63 | ||
| XM_007643740.1 | XP_007641930.1 | 10 - 11 | 1122 | 63 | ||
| XM_007643741.1 | XP_007641931.1 | 10 - 11 | 1190 | 63 | ||
| GenBank mRNA | JI895219.1 | - | 10 - 11 | 1149 | 63 | |
| JP055409.1 | - | 10 - 11 | 1149 | 63 | ||
Set Membership: |
|