Search
Search

Slc12a3| Gene Symbol: Slc12a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503611.2 | XP_003503659.1 | 24 - 25 | 2873 | 56 | |
| XM_007644200.1 | XP_007642390.1 | 25 - 26 | 2954 | 56 | ||
| XM_007644201.1 | XP_007642391.1 | 25 - 26 | 2951 | 56 | ||
| XM_007644202.1 | XP_007642392.1 | 24 - 25 | 2870 | 56 | ||
Set Membership: |
|