Search
Search

Klf8| Gene Symbol: Klf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648323.1 | XP_007646513.1 | 4 - 5 | 996 | 91 | |
| XM_007648324.1 | XP_007646514.1 | 4 - 5 | 476 | 91 | ||
| XM_007648325.1 | XP_007646515.1 | 3 - 4 | 817 | 91 | ||
Set Membership: |
|