Search
Search

Ebf3| Gene Symbol: Ebf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652419.1 | XP_007650609.1 | 8 - 9 | 748 | 66 | |
| XM_007652420.1 | XP_007650610.1 | 8 - 9 | 721 | 66 | ||
| XM_007652421.1 | XP_007650611.1 | 8 - 9 | 748 | 66 | ||
| XM_007652422.1 | XP_007650612.1 | 8 - 9 | 748 | 66 | ||
| XM_007652423.1 | XP_007650613.1 | 8 - 9 | 748 | 66 | ||
| XM_007652424.1 | XP_007650614.1 | 8 - 9 | 748 | 66 | ||
| XM_007652425.1 | XP_007650615.1 | 8 - 9 | 748 | 66 | ||
| XM_007652426.1 | XP_007650616.1 | 9 - 10 | 875 | 66 | ||
| XM_007652427.1 | XP_007650617.1 | 9 - 10 | 875 | 66 | ||
| XM_007652428.1 | XP_007650618.1 | 9 - 10 | 848 | 66 | ||
| XM_007652429.1 | XP_007650619.1 | 9 - 10 | 848 | 66 | ||
| XM_007652430.1 | XP_007650620.1 | 9 - 10 | 848 | 66 | ||
Set Membership: |
|