Search
Search

Ago2| Gene Symbol: Ago2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651571.1 | XP_007649761.1 | 15 - 16 | 2146 | 67 | |
| GenBank mRNA | JI891513.1 | - | 11 - 12 | 1492 | 67 | |
| JP049978.1 | - | 11 - 12 | 1491 | 67 | ||
Set Membership: |
|