Search
Search

Fasn| Gene Symbol: Fasn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496909.2 | XP_003496957.1 | 28 - 29 | 5158 | 55 | |
| GenBank mRNA | JI890764.1 | - | 18 - 19 | 3217 | 55 | |
| JP065322.1 | - | 18 - 19 | 3216 | 55 | ||
Set Membership: |
|