Search
Search

Vps45| Gene Symbol: Vps45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498635.2 | XP_003498683.1 | 10 - 11 | 1202 | 68 | |
| GenBank mRNA | JI878167.1 | - | 2 - 3 | 271 | 68 | |
| JP062534.1 | - | 2 - 3 | 271 | 68 | ||
Set Membership: |
|