Search
Search

Phf1| Gene Symbol: Phf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510828.2 | XP_003510876.1 | 11 - 12 | 1140 | 52 | |
| XM_007651408.1 | XP_007649598.1 | 11 - 12 | 1140 | 52 | ||
Set Membership: |
|