Search
Search

Hoxb6| Gene Symbol: Hoxb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499552.2 | XP_003499600.1 | 1 - 2 | 2838 | 61 | |
| XM_007640311.1 | XP_007638501.1 | 2 - 3 | 699 | 61 | ||
| XM_007640312.1 | XP_007638502.1 | 2 - 3 | 1542 | 61 | ||
| XM_007640313.1 | XP_007638503.1 | 2 - 3 | 630 | 61 | ||
Set Membership: |
|