Search
Search

Ltbp2| Gene Symbol: Ltbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652084.1 | XP_007650274.1 | 20 - 21 | 3629 | 56 | |
| XM_007652085.1 | XP_007650275.1 | 20 - 21 | 3629 | 56 | ||
Set Membership: |
|