Search
Search

Prpf40b| Gene Symbol: Prpf40b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513496.2 | XP_003513544.2 | 18 - 19 | 2301 | 83 | |
| XM_007653776.1 | XP_007651966.1 | 18 - 19 | 2301 | 83 | ||
| XM_007653777.1 | XP_007651967.1 | 18 - 19 | 2303 | 83 | ||
Set Membership: |
|