Search
Search

Usb1| Gene Symbol: Usb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509794.2 | XP_003509842.1 | 2 - 3 | 329 | 64 | |
| GenBank mRNA | JI869859.1 | - | 1 - 2 | 62 | 64 | |
| JP056074.1 | - | 1 - 2 | 62 | 64 | ||
Set Membership: |
|