Search
Search

Ccdc22| Gene Symbol: Ccdc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505079.2 | XP_003505127.1 | 12 - 13 | 1686 | 62 | |
| GenBank mRNA | JI868576.1 | - | 9 - 10 | 1166 | 62 | |
| JP059972.1 | - | 9 - 10 | 1166 | 62 | ||
Set Membership: |
|