Search
Search

Nfe2l1| Gene Symbol: Nfe2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499559.2 | XP_003499607.1 | 2 - 3 | 1109 | 100 | |
| GenBank mRNA | JI877268.1 | - | 1 - 2 | 79 | 99 | |
| JP047587.1 | - | 1 - 2 | 79 | 99 | ||
Set Membership: |
|