Search
Search

Bsdc1| Gene Symbol: Bsdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500633.2 | XP_003500681.1 | 5 - 6 | 444 | 60 | |
| XM_007641395.1 | XP_007639585.1 | 3 - 4 | 349 | 60 | ||
| GenBank mRNA | JI873084.1 | - | 5 - 6 | 444 | 60 | |
| JP062320.1 | - | 5 - 6 | 444 | 60 | ||
Set Membership: |
|